missing translation for 'onlineSavingsMsg'
Learn More

4-Metilamino-1-propionilpiperidina, 97 %, Thermo Scientific™

Catalog Number p-7033752
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
902836-34-4
C9H18N2O
170.26
MFCD08061056
RRUSDXAHDXKSFA-UHFFFAOYSA-N
4-methylamino-1-propionylpiperidine, 1-4-methylamino piperidin-1-yl propan-1-one, n-methyl-1-propionyl-4-piperidinamine, 1-4-methylamino piperidyl propan-1-one, 1-4-methylamino-piperidin-1-yl-propan-1-one
24208856
1-[4-(metilamino)piperidin-1-il]propan-1-ona
CCC(=O)N1CCC(CC1)NC
Este artículo no se puede devolver. Vea la política de devoluciones
902836-34-4
C9H18N2O
170.26
MFCD08061056
RRUSDXAHDXKSFA-UHFFFAOYSA-N
4-methylamino-1-propionylpiperidine, 1-4-methylamino piperidin-1-yl propan-1-one, n-methyl-1-propionyl-4-piperidinamine, 1-4-methylamino piperidyl propan-1-one, 1-4-methylamino-piperidin-1-yl-propan-1-one
24208856
1-[4-(metilamino)piperidin-1-il]propan-1-ona
CCC(=O)N1CCC(CC1)NC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Methylamino-1-propionylpiperidine
Cantidad 250 mg
Formula Weight (peso de la fórmula) 170.26
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'