Learn More
4-Methyl Hippuric Acid, TRC
Product Code. 30470160
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30470160 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30470160 Supplier Toronto Research Chemicals Supplier No. TRCM3119751G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methyl Hippuric Acid
CAS 27115-50-0
Purity Grade Notes HPLC
Molecular Formula C10 H11 N O3
Quantity 1 g
Synonym Glycine','N-(4-methylbenzoyl)-, Hippuric acid','p-methyl- (6CI,8CI), N-(4-Methylbenzoyl)glycine, (4-Methylbenzoylamino)acetic acid, 2-[(4-Methylbenzoyl)amino]acetic acid, 2-[(4-Methylphenyl)formamido]acetic acid, 4-Methylhippuric acid, N-(p-Methylbenzoyl)glycine, NSC 126814, [[(4-Methylphenyl)carbonyl]amino]acetic acid, p-Methylhippuric acid, p-Toluric acid, 4-Methyl hippuric acid
InChI Formula InChI=1S/C10H11NO3/c1-7-2-4-8(5-3-7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)
SMILES Cc1ccc(cc1)C(=O)NCC(=O)O
IUPAC Name 2-[(4-methylbenzoyl)amino]acetic acid
Molecular Weight (g/mol) 193.2
Formula Weight 193.0739
Percent Purity >95