missing translation for 'onlineSavingsMsg'
Learn More

4'-Metoxibifenil-4-carboxaldehído, 96 %, Thermo Scientific™

Catalog Number p-7031906
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
52988-34-8
C14H12O2
212.248
MFCD01862517
JTTIGLYPLMYHAT-UHFFFAOYSA-N
4-4-methoxyphenyl benzaldehyde, 4'-methoxy-1,1'-biphenyl-4-carbaldehyde, 4'-methoxybiphenyl-4-carbaldehyde, 4'-methoxy-biphenyl-4-carboxaldehyde, 4'-methoxy-biphenyl-4-carbaldehyde, 4'-methoxy 1,1'-biphenyl-4-carbaldehyde, 4'-methoxybiphenyl-4-carboxaldehyde, pubchem10292, methoxybiphenylcarbaldehyde, 4-4-methoxy benzaldehyde
1394487
4-(4-metoxifenil)benzaldehído
COC1=CC=C(C=C1)C2=CC=C(C=C2)C=O
Este artículo no se puede devolver. Vea la política de devoluciones
52988-34-8
C14H12O2
212.248
MFCD01862517
JTTIGLYPLMYHAT-UHFFFAOYSA-N
4-4-methoxyphenyl benzaldehyde, 4'-methoxy-1,1'-biphenyl-4-carbaldehyde, 4'-methoxybiphenyl-4-carbaldehyde, 4'-methoxy-biphenyl-4-carboxaldehyde, 4'-methoxy-biphenyl-4-carbaldehyde, 4'-methoxy 1,1'-biphenyl-4-carbaldehyde, 4'-methoxybiphenyl-4-carboxaldehyde, pubchem10292, methoxybiphenylcarbaldehyde, 4-4-methoxy benzaldehyde
1394487
4-(4-metoxifenil)benzaldehído
COC1=CC=C(C=C1)C2=CC=C(C=C2)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4'-Methoxybiphenyl-4-carboxaldehyde
Punto de fusión 98°C to 100°C
Temperatura de inflamación >110°C (230°F)
Cantidad 1 g
Sensibilidad Air Sensitive
Formula Weight (peso de la fórmula) 212.25
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'