missing translation for 'onlineSavingsMsg'
Learn More

Cloruro de (4-metoxibencilo)trifenilfosfonio, 97 %, Thermo Scientific Chemicals

Catalog Number p-7043398
Click to view available options
Cantidad:
10 g
50 g
Este artículo no se puede devolver. Vea la política de devoluciones
3462-97-3
C26H24ClOP
418.90
MFCD00031572
YQXBNCFNXOFWLR-UHFFFAOYSA-M
4-methoxybenzyl triphenylphosphonium chloride, 4-methoxyphenyl methyl triphenylphosphanium chloride, acmc-1ctv8, 4-methoxyphenyl methyl-triphenyl-phosphanium chloride, 4-methoxybenzyltriphenylphosphonium chloride, 4-methoxybenzyltriphenylphosphonium-chloride, 4-methoxybenzyl triphenyphosphonium chloride, p-methoxybenzyl-triphenylphosphonium chloride, 4-methoxybenzyltriphenyl phosphonium chloride, 4-methoxybenzyl triphenyl phosphonium chloride
2773651
(4-metoxifenil)metil-trifenilfosferio; cloruro
[Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
3462-97-3
C26H24ClOP
418.90
MFCD00031572
YQXBNCFNXOFWLR-UHFFFAOYSA-M
4-methoxybenzyl triphenylphosphonium chloride, 4-methoxyphenyl methyl triphenylphosphanium chloride, acmc-1ctv8, 4-methoxyphenyl methyl-triphenyl-phosphanium chloride, 4-methoxybenzyltriphenylphosphonium chloride, 4-methoxybenzyltriphenylphosphonium-chloride, 4-methoxybenzyl triphenyphosphonium chloride, p-methoxybenzyl-triphenylphosphonium chloride, 4-methoxybenzyltriphenyl phosphonium chloride, 4-methoxybenzyl triphenyl phosphonium chloride
2773651
(4-metoxifenil)metil-trifenilfosferio; cloruro
[Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (4-Methoxybenzyl)triphenylphosphonium chloride
Punto de fusión 235°C to 239°C
Cantidad 10 g
Beilstein 4173378
Formula Weight (peso de la fórmula) 418.91
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'