Learn More
4-Methoxy-3-[3-(morpholin-4-yl)propoxy]benzaldehyde, TRC
Product Code. 30466920
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30466920 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30466920 Supplier Toronto Research Chemicals Supplier No. TRCM152140500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methoxy-3-[3-(morpholin-4-yl)propoxy]benzaldehyde
CAS 861453-11-4
Molecular Formula C15 H21 N O4
Quantity 500 mg
Synonym Benzaldehyde, 4-methoxy-3-[3-(4-morpholinyl)propoxy]-, 4-Methoxy-3-[3-(4-morpholinyl)propoxy]benzaldehyde, 4-Methoxy-3-[3-(morpholin-4-yl)propoxy]benzaldehyde
InChI Formula InChI=1S/C15H21NO4/c1-18-14-4-3-13(12-17)11-15(14)20-8-2-5-16-6-9-19-10-7-16/h3-4,11-12H,2,5-10H2,1H3
SMILES COc1ccc(C=O)cc1OCCCN2CCOCC2
IUPAC Name 4-methoxy-3-(3-morpholin-4-ylpropoxy)benzaldehyde
Molecular Weight (g/mol) 279.33
Formula Weight 279.1471