Learn More
4-Hidroxi-2-(trifluorometil)piridina, 97 %, Thermo Scientific™
Catalog Number 15491559
Click to view available options
Cantidad:
250 mg
1 g
5 g
170886-13-2
C6H4F3NO
163.099
MFCD01571202
LKHCWMQMRXAPHX-UHFFFAOYSA-N
2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
10953849
2-(trifluorometil)-1H-piridin-4-ona
C1=CNC(=CC1=O)C(F)(F)F
170886-13-2
C6H4F3NO
163.099
MFCD01571202
LKHCWMQMRXAPHX-UHFFFAOYSA-N
2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
10953849
2-(trifluorometil)-1H-piridin-4-ona
C1=CNC(=CC1=O)C(F)(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Hydroxy-2-(trifluoromethyl)pyridine
Color Blanco
Cantidad 250 mg
Formula Weight (peso de la fórmula) 163.1
Porcentaje de pureza 97%
Forma física Polvo