missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-formil-3-(trifluorometil)bencenoborónico, 95 %, Thermo Scientific Chemicals

Catalog Number 15447645
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1072944-24-1
C8H6BF3O3
217.938
MFCD09037498
OQDHNEKEJVPVDS-UHFFFAOYSA-N
4-formyl-3-trifluoromethyl phenyl boronic acid, 4-formyl-3-trifluoromethyl phenylboronic acid, 4-formyl-3-trifluoromethyl benzeneboronic acid, 5-borono-2-formylbenzotrifluoride, 4-borono-2-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-formylphenylboronic acid, acmc-2098oi, 4-formyl-3-trifluoromethyl phenyl boronicacid
44755197
ácido [4-formil-3-(trifluorometil)fenil]borónico
B(C1=CC(=C(C=C1)C=O)C(F)(F)F)(O)O
Este artículo no se puede devolver. Vea la política de devoluciones
1072944-24-1
C8H6BF3O3
217.938
MFCD09037498
OQDHNEKEJVPVDS-UHFFFAOYSA-N
4-formyl-3-trifluoromethyl phenyl boronic acid, 4-formyl-3-trifluoromethyl phenylboronic acid, 4-formyl-3-trifluoromethyl benzeneboronic acid, 5-borono-2-formylbenzotrifluoride, 4-borono-2-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-formylphenylboronic acid, acmc-2098oi, 4-formyl-3-trifluoromethyl phenyl boronicacid
44755197
ácido [4-formil-3-(trifluorometil)fenil]borónico
B(C1=CC(=C(C=C1)C=O)C(F)(F)F)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Formyl-3-(trifluoromethyl)benzeneboronic acid
Punto de fusión 200°C to 202°C
Cantidad 250 mg
Sensibilidad Air Sensitive
Formula Weight (peso de la fórmula) 217.94
Porcentaje de pureza 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'