missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-fluoro-2-(trifluorometoxi)benzoico, 97 %, Thermo Scientific™

Catalog Number p-7032043
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1242250-37-8
C8H4F4O3
224.111
MFCD19687173
GSEJXXUTVPFRRV-UHFFFAOYSA-N
4-fluoro-2-trifluoromethoxy benzoic acid
66523582
ácido 4-fluoro-2-(trifluorometoxi)benzoico
C1=CC(=C(C=C1F)OC(F)(F)F)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
1242250-37-8
C8H4F4O3
224.111
MFCD19687173
GSEJXXUTVPFRRV-UHFFFAOYSA-N
4-fluoro-2-trifluoromethoxy benzoic acid
66523582
ácido 4-fluoro-2-(trifluorometoxi)benzoico
C1=CC(=C(C=C1F)OC(F)(F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Fluoro-2-(trifluoromethoxy)benzoic acid
Cantidad 1 g
Formula Weight (peso de la fórmula) 224.11
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'