missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluorometoxi)benzamida, 97 %

Catalog Number p-7032151
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1323966-41-1
C8H5F4NO2
223.13
MFCD18907236
MNJBQYPDEXYTJF-UHFFFAOYSA-N
4-fluoro-2-trifluoromethoxy benzamide
66523581
4-fluoro-2-(trifluorometoxi)benzamida
NC(=O)C1=C(OC(F)(F)F)C=C(F)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
1323966-41-1
C8H5F4NO2
223.13
MFCD18907236
MNJBQYPDEXYTJF-UHFFFAOYSA-N
4-fluoro-2-trifluoromethoxy benzamide
66523581
4-fluoro-2-(trifluorometoxi)benzamida
NC(=O)C1=C(OC(F)(F)F)C=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Fluoro-2-(trifluoromethoxy)benzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 223.12
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'