missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-fluoro-2-isopropoxibencenoborónico, 97 %, Thermo Scientific™

Catalog Number p-7052692
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
480438-59-3
C9H12BFO3
198
MFCD06411287
RSEFJQODCQCGRB-UHFFFAOYSA-N
4-fluoro-2-isopropoxyphenylboronic acid, 4-fluoro-2-isopropoxyphenyl boronic acid, 4-fluoro-2-propan-2-yloxy phenyl boronic acid, 4-fluoro-2-isopropoxybenzeneboronic acid, 4-fluoro-2-prop-2-yl oxy benzeneboronic acid, acmc-1apz6, 4-fluoro-2-isopropoxyphenylboronicacid, 4-fluoro-2-isopropoxyphenyl boronicacid, 4-fluoro-2-isopropoxybenzene boronic acid, 4-fluoro-2-propan-2-yl oxy phenyl boronic acid
16217202
ácido (4-fluoro-2-propan-2-iloxyfenil)borónico
B(C1=C(C=C(C=C1)F)OC(C)C)(O)O
Este artículo no se puede devolver. Vea la política de devoluciones
480438-59-3
C9H12BFO3
198
MFCD06411287
RSEFJQODCQCGRB-UHFFFAOYSA-N
4-fluoro-2-isopropoxyphenylboronic acid, 4-fluoro-2-isopropoxyphenyl boronic acid, 4-fluoro-2-propan-2-yloxy phenyl boronic acid, 4-fluoro-2-isopropoxybenzeneboronic acid, 4-fluoro-2-prop-2-yl oxy benzeneboronic acid, acmc-1apz6, 4-fluoro-2-isopropoxyphenylboronicacid, 4-fluoro-2-isopropoxyphenyl boronicacid, 4-fluoro-2-isopropoxybenzene boronic acid, 4-fluoro-2-propan-2-yl oxy phenyl boronic acid
16217202
ácido (4-fluoro-2-propan-2-iloxyfenil)borónico
B(C1=C(C=C(C=C1)F)OC(C)C)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Fluoro-2-isopropoxybenzeneboronic acid
Cantidad 250 mg
Formula Weight (peso de la fórmula) 198
Porcentaje de pureza 97%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'