Learn More
4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Product Code. 11926101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11926101 5 g 5g
11916101 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926101 Supplier Thermo Scientific Acros Supplier No. 445000050
 Restricted Item
Fecha estimada de envoi: 11-12-2026
Add to basket
This item is not returnable. View return policy
CAS: 859169-20-3 | Molecular Formula: C16H23BO4 | Molecular Weight: 290.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 859169-20-3
Molecular Formula C16H23BO4
Molecular Weight (g/mol) 290.17
MDL Number MFCD08704684
InChI Key RPRBNRDIESZHFL-UHFFFAOYSA-N
Synonym ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester
PubChem CID 46737998
IUPAC Name ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Specifications
Melting Point 41°C to 43°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 290.16
Percent Purity 97%
Chemical Name or Material 4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.