missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopyridine-2-carboxylic acid, 95%

Product Code. 15467167
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467167 1 g 1g
15477167 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467167 Supplier Thermo Scientific Alfa Aesar Supplier No. H34233.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 640296-19-1 | C7H4N2O2 | 148.121 g/mol

4-cyanopyridine, and p- chlorobenzonitrile system which produces only amides, without the corresponding carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-cyanopyridine, and p- chlorobenzonitrile system which produces only amides, without the corresponding carboxylic acids.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 640296-19-1
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD09152773
InChI Key CMKZSIGRFONKKC-UHFFFAOYSA-N
Synonym 4-cyano-2-pyridinecarboxylic acid, 4-cyanopicolinic acid, 2-pyridinecarboxylic acid, 4-cyano, 4-cyano-2-picolinic acid, 4-cyano-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 4-cyano-9ci
PubChem CID 24213769
IUPAC Name 4-cyanopyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1C#N)C(=O)O

Specifications

Melting Point ∽210°C (decomposition)
Quantity 1 g
UN Number UN3439
Formula Weight 148.1
Percent Purity 95%
Chemical Name or Material 4-Cyanopyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.