missing translation for 'onlineSavingsMsg'
Learn More

(4-clorofenilsulfonil)acetona, 98 %, Thermo Scientific™

Catalog Number p-7022207
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
5000-48-6
C9H9ClO3S
232.68
MFCD00018663
BRDBHPZILGTBFY-UHFFFAOYSA-N
4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
521269
1-(4-clorofenil)sulfonilpropian-2-ona
CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
5000-48-6
C9H9ClO3S
232.68
MFCD00018663
BRDBHPZILGTBFY-UHFFFAOYSA-N
4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
521269
1-(4-clorofenil)sulfonilpropian-2-ona
CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (4-Chlorophenylsulfonyl)acetone
Punto de fusión 78°C to 82°C
Cantidad 1 g
Beilstein 2051464
Formula Weight (peso de la fórmula) 232.69
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'