missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-clorocinámico, predominantemente trans, 99 %, Thermo Scientific Chemicals

Catalog Number p-7022490
Click to view available options
Cantidad:
25 g
100 g
500 g
1615-02-7
C9H7ClO2
182.60
MFCD00004396
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
637797
CHEBI:61116
ácido (E)-3-(4-clorofenil)prop-2-enoico
OC(=O)\C=C\C1=CC=C(Cl)C=C1
1615-02-7
C9H7ClO2
182.60
MFCD00004396
GXLIFJYFGMHYDY-ZZXKWVIFSA-N
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
637797
CHEBI:61116
ácido (E)-3-(4-clorofenil)prop-2-enoico
OC(=O)\C=C\C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Chlorocinnamic acid
Nombre de nota predominantly trans
Punto de fusión 248°C to 253°C
Cantidad 25 g
Beilstein 1365308
Formula Weight (peso de la fórmula) 182.61
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'