missing translation for 'onlineSavingsMsg'
Learn More

4-Cloro-n,n-dimetil-2-nitroanilina, 97 %, Thermo Scientific™

Catalog Number 15489708
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
17815-99-5
C8H9ClN2O2
200.622
QOMAYFFYZHAPEH-UHFFFAOYSA-N
4-chloro-2-nitro-phenyl-dimethyl-amine, 2-nitro-4-chloro-n,n-dimethylaniline, 4-chloro-n,n-dimethyl-2-nitrobenzenamine, benzenamine, 4-chloro-n,n-dimethyl-2-nitro
12468660
4-cloro-n,n-dimetil-2-nitroanilina
CN(C)C1=C(C=C(C=C1)Cl)[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
17815-99-5
C8H9ClN2O2
200.622
QOMAYFFYZHAPEH-UHFFFAOYSA-N
4-chloro-2-nitro-phenyl-dimethyl-amine, 2-nitro-4-chloro-n,n-dimethylaniline, 4-chloro-n,n-dimethyl-2-nitrobenzenamine, benzenamine, 4-chloro-n,n-dimethyl-2-nitro
12468660
4-cloro-n,n-dimetil-2-nitroanilina
CN(C)C1=C(C=C(C=C1)Cl)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Chloro-N,N-dimethyl-2-nitroaniline
Cantidad 1 g
Número UN UN2811
Beilstein 2213297
Formula Weight (peso de la fórmula) 200.62
Porcentaje de pureza 97%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'