Learn More
4-Cloro-N-(2,6-diclorofenil)benzamida, 97 %, Thermo Scientific™
Catalog Number p-7039690
Click to view available options
Cantidad:
250 mg
1 g
200807-46-1
C13H8Cl3NO
300.563
MFCD00028788
MGCCUXCCTBKJAN-UHFFFAOYSA-N
4-chloro-n-2,6-dichlorophenyl benzamide, 2',4,6'-trichlorobenzanilide
4693001
4-cloro-N-(2,6-diclorofenil)benzamida
C1=CC(=C(C(=C1)Cl)NC(=O)C2=CC=C(C=C2)Cl)Cl
200807-46-1
C13H8Cl3NO
300.563
MFCD00028788
MGCCUXCCTBKJAN-UHFFFAOYSA-N
4-chloro-n-2,6-dichlorophenyl benzamide, 2',4,6'-trichlorobenzanilide
4693001
4-cloro-N-(2,6-diclorofenil)benzamida
C1=CC(=C(C(=C1)Cl)NC(=O)C2=CC=C(C=C2)Cl)Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Chloro-N-(2,6-dichlorophenyl)benzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 300.57
Porcentaje de pureza 97%