missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™

Product Code. 11450641
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450641 5 g 5g
11440641 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450641 Supplier Thermo Scientific Alfa Aesar Supplier No. B20119.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1735-54-2
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.564
MDL Number MFCD00190117
InChI Key VBPYHJRQZZCFCW-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzonitrile, 2-chloro-5-cyanobenzotrifluoride, 3-trifluoromethyl-4-chlorobenzonitrile, benzonitrile, 4-chloro-3-trifluoromethyl, 4-chloro-3-trifluoromethyl benzenecarbonitrile, pubchem8192, ksc494o5b, 4-chloro-3-trifluoromethyl-benzonitrile
PubChem CID 2736474
IUPAC Name 4-chloro-3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)C(F)(F)F)Cl

Specifications

Melting Point 64°C to 68°C
Boiling Point 100°C to 105°C (11 mmHg)
Odor Characteristic
Quantity 5 g
UN Number UN3439
Beilstein 2103768
Formula Weight 205.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.