Learn More
Integra Biosciences™ 4-Channel Adapter
Product Code. 10563642
Change view
Click to view available options
Type:
4-channel Adapter
Unit Size:
Each
Product Code. Type Unit Size
10563642 4-channel Adapter Each
1 options
This item is not returnable. View return policy
Product Code. 10563642 Supplier Integra Biosciences™ Supplier No. 155524
  • ONLINE EXCLUSIVE PRICE
    63.10€
    49.85€ / paquete de 1
    Ahorro 13.25€
    20% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
Includes Stainless Steel Tips
Length (Metric) Tip 40 mm
Type 4-channel Adapter