missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-bromo-2-cloro-5-fluorobenzoico, 98 %, Thermo Scientific™

Catalog Number p-7055396
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
177480-81-8
C7H3BrClFO2
253.45
MFCD00236589
TZFKKLGCVKORLS-UHFFFAOYSA-N
4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
2736225
4-bromo-2-chloro-5-fluorobenzoic acid
OC(=O)C1=CC(F)=C(Br)C=C1Cl
Este artículo no se puede devolver. Vea la política de devoluciones
177480-81-8
C7H3BrClFO2
253.45
MFCD00236589
TZFKKLGCVKORLS-UHFFFAOYSA-N
4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
2736225
4-bromo-2-chloro-5-fluorobenzoic acid
OC(=O)C1=CC(F)=C(Br)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Bromo-2-chloro-5-fluorobenzoic acid
Color Blanco
Cantidad 1 g
Formula Weight (peso de la fórmula) 253.45
Porcentaje de pureza 98%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'