Learn More
4-Bromo-2,5-dimethoxybenzaldehyde, TRC
Product Code. 30345884
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30345884 5 g 5g
1 options
This item is not returnable. View return policy
Product Code. 30345884 Supplier Toronto Research Chemicals Supplier No. TRCB6835455G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Bromo-2,5-dimethoxybenzaldehyde
CAS 31558-41-5
Molecular Formula C9 H9 Br O3
Quantity 5 g
Synonym 2,5-Dimethoxy-4-bromobenzaldehyde
InChI Formula InChI=1S/C9H9BrO3/c1-12-8-4-7(10)9(13-2)3-6(8)5-11/h3-5H,1-2H3
SMILES COc1cc(C=O)c(OC)cc1Br
IUPAC Name 4-bromo-2,5-dimethoxybenzaldehyde
Molecular Weight (g/mol) 245.07
Formula Weight 243.974