missing translation for 'onlineSavingsMsg'
Learn More

4-Benciloxifenilacetonitrilo, 97 %

Catalog Number p-7045195
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
838-96-0
C15H13NO
223.28
MFCD00016400
QKEYZRVDFZDOEP-UHFFFAOYSA-N
4-benzyloxyphenylacetonitrile, 2-4-benzyloxy phenyl acetonitrile, 4-benzyloxybenzyl cyanide, 4-benzyloxy phenyl acetonitrile, 2-4-phenylmethoxy phenyl ethanenitrile, acmc-20amoi, pubchem15926, 4-benzyloxybenzeneacetonitrile, p-benzyloxyphenyl acetonitrile
522804
2-(4-fenilmetoxifenilo)acetonitrilo
N#CCC1=CC=C(OCC2=CC=CC=C2)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
838-96-0
C15H13NO
223.28
MFCD00016400
QKEYZRVDFZDOEP-UHFFFAOYSA-N
4-benzyloxyphenylacetonitrile, 2-4-benzyloxy phenyl acetonitrile, 4-benzyloxybenzyl cyanide, 4-benzyloxy phenyl acetonitrile, 2-4-phenylmethoxy phenyl ethanenitrile, acmc-20amoi, pubchem15926, 4-benzyloxybenzeneacetonitrile, p-benzyloxyphenyl acetonitrile
522804
2-(4-fenilmetoxifenilo)acetonitrilo
N#CCC1=CC=C(OCC2=CC=CC=C2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Benzyloxyphenylacetonitrile
Punto de fusión 66°C to 70°C
Punto de ebullición 170°C to 173°C (0.1 mmHg)
Cantidad 250 mg
Beilstein 2377584
Formula Weight (peso de la fórmula) 223.28
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'