Learn More
Éster de pinacol del ácido 4-amino-2-cloro-3-fluorobencenoborónico, 96 %, Thermo Scientific™
Catalog Number p-7054494
Click to view available options
Cantidad:
250 mg
1 g
1154740-65-4
C12H16BClFNO2
271.52
MFCD16996254
NFNKJOVOJSFUSA-UHFFFAOYSA-N
3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
59606745
3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1
1154740-65-4
C12H16BClFNO2
271.52
MFCD16996254
NFNKJOVOJSFUSA-UHFFFAOYSA-N
3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-amino-2-chloro-3-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 3-chloro-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
59606745
3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notas
Almacenar en lugar fresco y seco en un recipiente cerrado herméticamente. Garantice una ventilación adecuada. Almacenar alejado de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester
Color Blanco
Cantidad 250 mg
Formula Weight (peso de la fórmula) 271.52
Porcentaje de pureza 96%
Forma física Sólido