Learn More
Cloruro de 4,5,6,7-tetrahidrobenzo[b]tiofeno-2-sulfonilo, 95 %, Thermo Scientific Chemicals
Catalog Number p-7032839
Click to view available options
Cantidad:
250 mg
1 g
128852-17-5
C8H9ClO2S2
236.73
MFCD18825753
UUBSHLWNADKCBU-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci
15158650
4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
ClS(=O)(=O)C1=CC2=C(CCCC2)S1
128852-17-5
C8H9ClO2S2
236.73
MFCD18825753
UUBSHLWNADKCBU-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci
15158650
4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
ClS(=O)(=O)C1=CC2=C(CCCC2)S1
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El cloruro de 4,5,6,7-tetrahidrobenzo[b]tiofeno-2-sulfonilo se utiliza como intermedio farmacéutico.
Solubilidad
Ligeramente soluble en agua
Notas
Sensible a la humedad. Incompatible con los agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride
Punto de fusión 64°C to 66°C
Cantidad 250 mg
Número UN UN3261
Sensibilidad Moisture sensitive
Información de solubilidad Slightly soluble in water
Formula Weight (peso de la fórmula) 236.74
Porcentaje de pureza 95%