Learn More
4-[4-(Trifluoromethoxy)phenoxy]piperidine, TRC
Product Code. 30449748
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30449748 5 g 5g
1 options
This item is not returnable. View return policy
Product Code. 30449748 Supplier Toronto Research Chemicals Supplier No. TRCT7908905G
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine
CAS 287952-67-4
Molecular Formula C12H14F3NO2
Quantity 5 g
InChI Formula InChI=1S/C12H14F3NO2/c13-12(14,15)18-11-3-1-9(2-4-11)17-10-5-7-16-8-6-10/h1-4,10,16H,5-8H2
SMILES FC(F)(F)Oc1ccc(OC2CCNCC2)cc1
IUPAC Name 4-[4-(trifluoromethoxy)phenoxy]piperidine
Molecular Weight (g/mol) 261.24
Formula Weight 261.0977