missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-(4'-metoxi-4-bifenilsulfonilamino)benzoico, 96 %, Thermo Scientific™

Catalog Number p-7031674
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
885269-42-1
C20H17NO5S
383.418
MFCD06409458
ZFNQOUZSNZLJLR-UHFFFAOYSA-N
4-4'-methoxy-1,1'-biphenyl-4-sulfonamido benzoic acid, 4-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido benzoic acid, 4-4'-methoxy-4-biphenylylsulfonylamino benzoic acid
44669168
ácido 4-[[4-(4-metoxifenil)fenil]sulfonilamino]benzoico
COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
885269-42-1
C20H17NO5S
383.418
MFCD06409458
ZFNQOUZSNZLJLR-UHFFFAOYSA-N
4-4'-methoxy-1,1'-biphenyl-4-sulfonamido benzoic acid, 4-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido benzoic acid, 4-4'-methoxy-4-biphenylylsulfonylamino benzoic acid
44669168
ácido 4-[[4-(4-metoxifenil)fenil]sulfonilamino]benzoico
COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(4'-Methoxy-4-biphenylylsulfonylamino)benzoic acid
Punto de fusión >250°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 383.43
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'