Learn More
Alfa Aesar™ 4-(4-Fluorobenzyloxy)benzyl alcohol, 98%
Marca: Alfa Aesar™ 044022.14
Detalles adicionales : CAS : 117113-98-1 Peso : 0.02500kg
Cantidad. | 25g |
---|
Identificadores de productos químicos
117113-98-1 | |
232.254 | |
FNWSEJASZBRBDB-UHFFFAOYSA-N | |
2737278 | |
C1=CC(=CC=C1CO)OCC2=CC=C(C=C2)F |
C14H13FO2 | |
MFCD00173648 | |
4-4-fluorobenzyloxy benzyl alcohol, 4-4-fluorobenzyl oxy phenyl methanol, 4-4-fluorophenyl methoxy phenyl methanol, 4-4-fluorophenyl methoxy phenyl methanol, 4-4-fluorobenzyloxy benzylalcohol, 4-4-fluorobenzyl oxy phenyl methanol, 4-4-fluorophenyl methoxy phenyl methan-1-ol, maybridge3_000507, acmc-1bo74, 4-4'-fluorobenzyloxy benzyl alcohol | |
[4-[(4-fluorophenyl)methoxy]phenyl]methanol |
Especificación
4- (4-Fluorobenzyloxy)benzyl alcohol | |
117113-98-1 | |
MFCD00173648 | |
FNWSEJASZBRBDB-UHFFFAOYSA-N | |
[4-[(4-fluorophenyl)methoxy]phenyl]methanol | |
2737278 | |
Powder |
25g | |
C14H13FO2 | |
4-4-fluorobenzyloxy benzyl alcohol, 4-4-fluorobenzyl oxy phenyl methanol, 4-4-fluorophenyl methoxy phenyl methanol, 4-4-fluorophenyl methoxy phenyl methanol, 4-4-fluorobenzyloxy benzylalcohol, 4-4-fluorobenzyl oxy phenyl methanol, 4-4-fluorophenyl methoxy phenyl methan-1-ol, maybridge3_000507, acmc-1bo74, 4-4'-fluorobenzyloxy benzyl alcohol | |
C1=CC(=CC=C1CO)OCC2=CC=C(C=C2)F | |
232.254 | |
232.26 | |
98% |