missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluoro-2-nitrofenil)morfolina, 97 %, Thermo Scientific™

Catalog Number 15409848
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
238418-75-2
C10H11FN2O3
226.21
MFCD00798369
XEVFYVWDTJTEHQ-UHFFFAOYSA-N
4-4-fluoro-2-nitrophenyl morpholine, acmc-1cbj8, 5-fluoro-2-morpholinonitrobenzene, 4-4-fluoro-2-nitro-phenyl-morpholine, morpholine,4-4-fluoro-2-nitrophenyl
2737431
4-(4-fluoro-2-nitrophenyl)morpholine
[O-][N+](=O)C1=C(C=CC(F)=C1)N1CCOCC1
Este artículo no se puede devolver. Vea la política de devoluciones
238418-75-2
C10H11FN2O3
226.21
MFCD00798369
XEVFYVWDTJTEHQ-UHFFFAOYSA-N
4-4-fluoro-2-nitrophenyl morpholine, acmc-1cbj8, 5-fluoro-2-morpholinonitrobenzene, 4-4-fluoro-2-nitro-phenyl-morpholine, morpholine,4-4-fluoro-2-nitrophenyl
2737431
4-(4-fluoro-2-nitrophenyl)morpholine
[O-][N+](=O)C1=C(C=CC(F)=C1)N1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(4-Fluoro-2-nitrophenyl)morpholine
Cantidad 25 g
Formula Weight (peso de la fórmula) 226.2
Porcentaje de pureza 97%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'