Learn More
4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid), Mikromol™
Product Code. 30050270
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050270 100 mg 100mg
30050220 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050270 Supplier Mikromol Supplier No. MM0918.05
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid)
CAS 3984-34-7
Analyte or Component Names 4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid)
Shipping Condition Room Temperature
Molecular Formula C10 H9 Cl O3
Certifications/Compliance ISO 17025
Synonym 4-(4-Chlorophenyl)-4-oxobutanoic Acid, 3-(4-Chlorobenzoyl)propionic Acid
InChI Formula InChI=1S/C10H9ClO3/c11-8-3-1-7(2-4-8)9(12)5-6-10(13)14/h1-4H,5-6H2,(H,13,14)
SMILES OC(=O)CCC(=O)c1ccc(Cl)cc1
IUPAC Name 4-(4-chlorophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol) 212.63
Quantity 100 mg
Formula Weight 212.024 g/mol