missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(2,5-dimethylstyryl)biphenyl, 99+%, Thermo Scientific™

Catalog Number p-7045034
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
72814-85-8
C32H30
414.59
MFCD00051223
AMDSCXLLKXHWOJ-IWGRKNQJSA-N
4,4'-bis 2,5-dimethylstyryl-1,1'-biphenyl, 1,1'-biphenyl, 4,4'-bis 2-2,5-dimethylphenyl ethenyl, 4,4'-bis 2,5-dimethylstyryl biphenyl, 4,4'-bis-2,5-dimethylstyryl biphenyl, 4,4'-bis e-2-2,5-dimethylphenyl ethenyl-1,1'-biphenyl, 2-1e-2-4-4-1e-2-2,5-dimethylphenyl vinyl phenyl phenyl vinyl-1,4-dim ethylbenzene, 2-e-2-4-4-e-2-2,5-dimethylphenyl ethenyl phenyl phenyl ethenyl-1,4-dimethylbenzene
6442034
2-[(E)-2-[4-[4-[(E)-2-(2,5-dimetil fenil)etenil]fenil]fenil]etenil]-1,4-dimetilbenceno
CC1=CC=C(C)C(\C=C\C2=CC=C(C=C2)C2=CC=C(\C=C\C3=CC(C)=CC=C3C)C=C2)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
72814-85-8
C32H30
414.59
MFCD00051223
AMDSCXLLKXHWOJ-IWGRKNQJSA-N
4,4'-bis 2,5-dimethylstyryl-1,1'-biphenyl, 1,1'-biphenyl, 4,4'-bis 2-2,5-dimethylphenyl ethenyl, 4,4'-bis 2,5-dimethylstyryl biphenyl, 4,4'-bis-2,5-dimethylstyryl biphenyl, 4,4'-bis e-2-2,5-dimethylphenyl ethenyl-1,1'-biphenyl, 2-1e-2-4-4-1e-2-2,5-dimethylphenyl vinyl phenyl phenyl vinyl-1,4-dim ethylbenzene, 2-e-2-4-4-e-2-2,5-dimethylphenyl ethenyl phenyl phenyl ethenyl-1,4-dimethylbenzene
6442034
2-[(E)-2-[4-[4-[(E)-2-(2,5-dimetil fenil)etenil]fenil]fenil]etenil]-1,4-dimetilbenceno
CC1=CC=C(C)C(\C=C\C2=CC=C(C=C2)C2=CC=C(\C=C\C3=CC(C)=CC=C3C)C=C2)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4,4'-Bis(2,5-dimethylstyryl)biphenyl
Punto de fusión 196°C to 200°C
Beilstein 6526037
Formula Weight (peso de la fórmula) 414.59
Porcentaje de pureza ≥99%
Cantidad 1 g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'