missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 4-(3-etoxicarbonil-1-piperidinilcarbonil) bencenoborónico, 98 %

Catalog Number p-7030348
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
850411-14-2
C21H30BNO5
387.283
MFCD06657751
YQADGHATUTVIEA-UHFFFAOYSA-N
4-3-ethoxycarbonylpiperidine carboxamidophenylboronic acid, pinacol ester, ethyl 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperidine-3-carboxylate, 4-3-ethoxycarbonyl-1-piperidinylcarbonyl benzeneboronic acid pinacol ester, 4-ethyl-3'-piperidinecarboxylate-1-carbonyl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoyl-piperidine-3-carboxylic acid ethyl ester, amtb023, 4-3-ethoxycarbonyl-1-piperidinylcarbonyl benzeneboronicacidpinacolester, 4-3-ethoxycarbonylpiperidine carboxamidophenylboronic acid pinacol ester
17998917
1-[4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)benzoil]piperidina-3-carboxilato de etilo
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)N3CCCC(C3)C(=O)OCC
Este artículo no se puede devolver. Vea la política de devoluciones
850411-14-2
C21H30BNO5
387.283
MFCD06657751
YQADGHATUTVIEA-UHFFFAOYSA-N
4-3-ethoxycarbonylpiperidine carboxamidophenylboronic acid, pinacol ester, ethyl 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperidine-3-carboxylate, 4-3-ethoxycarbonyl-1-piperidinylcarbonyl benzeneboronic acid pinacol ester, 4-ethyl-3'-piperidinecarboxylate-1-carbonyl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoyl-piperidine-3-carboxylic acid ethyl ester, amtb023, 4-3-ethoxycarbonyl-1-piperidinylcarbonyl benzeneboronicacidpinacolester, 4-3-ethoxycarbonylpiperidine carboxamidophenylboronic acid pinacol ester
17998917
1-[4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)benzoil]piperidina-3-carboxilato de etilo
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)N3CCCC(C3)C(=O)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 4-(3-Ethoxycarbonyl-1-piperidinylcarbonyl)benzeneboronic acid pinacol ester
Cantidad 1 g
Formula Weight (peso de la fórmula) 387.28
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'