Learn More
[(3S)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride ((1R,3S)-Benazepril Hydrochloride), Mikromol™
Product Code. 30051127
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30051127 25 mg 25mg
1 options
This item is not returnable. View return policy
Product Code. 30051127 Supplier Mikromol Supplier No. MM1229.080025
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[(3S)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride ((1R,3S)-Benazepril Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (3S)-3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid, monohydrochloride
CAS 86541-77-7
Analyte or Component Names (3S)-3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid, monohydrochloride
Shipping Condition Room Temperature
Molecular Formula C24 H28 N2 O5 . Cl H
Certifications/Compliance ISO 17025
Synonym Benazepril Hydrochloride Imp. B (EP), Benazepril Imp. B (EP), Benazepril USP Related Compound B, Benazepril USP RC B, (3S)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic Acid Hydrochloride (1:1), 1H-1-Benzazepine-1-acetic acid, 3-[[(1R)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, monohydrochloride, (3S)- (9CI), 1H-1-Benzazepine-1-acetic acid, 3-[[1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, monohydrochloride, [S-(R*,S*)]-, Benazepril Related Compound B as Hydrochloride, Benazepril Hydrochloride Impurity B as (1R,3S)-Enantiomer Hydrochloride, Benazepril Impurity B as (1R,3S)-Enantiomer Hydrochloride
InChI Formula InChI=1S/C24H28N2O5.ClH/c1-2-31-24(30)20(14-12-17-8-4-3-5-9-17)25-19-15-13-18-10-6-7-11-21(18)26(23(19)29)16-22(27)28;/h3-11,19-20,25H,2,12-16H2,1H3,(H,27,28);1H/t19-,20+;/m0./s1
SMILES Cl.CCOC(=O)[C@@H](CCc1ccccc1)N[C@H]2CCc3ccccc3N(CC(=O)O)C2=O
IUPAC Name 2-[(3S)-3-[[(2R)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetic acid;hydrochloride
Molecular Weight (g/mol) 460.95
Quantity 25 mg
Formula Weight 460.1765 g/mol