Learn More
3M™ Versaflo™ M-300 Series PAPR Replacement Head Suspension
Product Code. 15720078 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-300 Series Hardhats and M-400 Series Helmets
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15720078 M-300 Series Hardhats and M-400 Series Helmets Each
1 options
This item is not returnable. View return policy
Product Code. 15720078 Supplier 3M™ Supplier No. 7100063895
  • ONLINE EXCLUSIVE PRICE
    94.00€
    65.29€ / paquete de 1
    Ahorro 28.71€
    30% Descuento
Fecha estimada de envoi: 14-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Head suspension is a replacement part for Versaflo™ M-300 Series hardhats and M-400 Series helmets.
  • 3M Catalog# M-350
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Product Type Head Suspension
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-300 Series Hardhats and M-400 Series Helmets
Model M-350