missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluorometil)bencenosulfonamida, 97 %, Thermo Scientific™

Catalog Number p-7047755
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
672-58-2
C7H6F3NO2S
225.185
MFCD00042421
ZUTVRDMZQSHCID-UHFFFAOYSA-N
3-trifluoromethyl benzenesulfonamide, 3-trifluoromethyl benzene-1-sulfonamide, 3-trifluoromethyl benzenesulphonamide, m-trifluoromethylbenzenesulfonamide, benzenesulfonamide, 3-trifluoromethyl, pubchem11763, maybridge3_000947, 3-sulphamoylbenzotrifluoride, ksc493o8t, 3-trifluoromethyl-benzenesulfonamide
244076
3-(trifluorometil)bencenosulfonamida
C1=CC(=CC(=C1)S(=O)(=O)N)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
672-58-2
C7H6F3NO2S
225.185
MFCD00042421
ZUTVRDMZQSHCID-UHFFFAOYSA-N
3-trifluoromethyl benzenesulfonamide, 3-trifluoromethyl benzene-1-sulfonamide, 3-trifluoromethyl benzenesulphonamide, m-trifluoromethylbenzenesulfonamide, benzenesulfonamide, 3-trifluoromethyl, pubchem11763, maybridge3_000947, 3-sulphamoylbenzotrifluoride, ksc493o8t, 3-trifluoromethyl-benzenesulfonamide
244076
3-(trifluorometil)bencenosulfonamida
C1=CC(=CC(=C1)S(=O)(=O)N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(Trifluoromethyl)benzenesulfonamide
Punto de fusión 122°C to 126°C
Cantidad 1 g
Beilstein 2696480
Formula Weight (peso de la fórmula) 225.19
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'