missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3-(trifluorometoxi)benzoico, 99 %, Thermo Scientific™

Catalog Number p-7023992
Click to view available options
Cantidad:
1 g
5 g
25 g
This item is not returnable. View return policy
1014-81-9
C8H5F3O3
206.12
MFCD00041501
OKPFIWIMBJNFSE-UHFFFAOYSA-N
3-trifluoromethoxy benzoic acid, m-trifluoromethoxybenzoic acid, m-trifluoromethoxy benzoic acid, benzoic acid, 3-trifluoromethoxy, 3-trifluoromethoxy benzoicacid, alpha,alpha,alpha-trifluoro-m-anisic acid, 3-carboxy-alpha,alpha,alpha-trifluoroanisole, 3-trifluoromethoxy-benzoic acid, pubchem4590, acmc-1btyb
66096
ácido 3-(trifluorometoxi)benzoico
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O
This item is not returnable. View return policy
1014-81-9
C8H5F3O3
206.12
MFCD00041501
OKPFIWIMBJNFSE-UHFFFAOYSA-N
3-trifluoromethoxy benzoic acid, m-trifluoromethoxybenzoic acid, m-trifluoromethoxy benzoic acid, benzoic acid, 3-trifluoromethoxy, 3-trifluoromethoxy benzoicacid, alpha,alpha,alpha-trifluoro-m-anisic acid, 3-carboxy-alpha,alpha,alpha-trifluoroanisole, 3-trifluoromethoxy-benzoic acid, pubchem4590, acmc-1btyb
66096
ácido 3-(trifluorometoxi)benzoico
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(Trifluoromethoxy)benzoic acid
Punto de fusión 88°C to 89°C
Cantidad 1 g
Beilstein 2579574
Formula Weight (peso de la fórmula) 206.12
Porcentaje de pureza 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'