missing translation for 'onlineSavingsMsg'
Learn More

Clorhidrato de 3-quinuclidinona, +98 %, Thermo Scientific Chemicals

Catalog Number p-7021118
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
1193-65-3
C7H12ClNO
161.629
MFCD00137391
RFDPHKHXPMDJJD-UHFFFAOYSA-N
3-quinuclidinone hydrochloride, quinuclidin-3-one hydrochloride, 3-quinuclidone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one hydrochloride, 3-quinuclidnone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride, 1-aza-3-oxobicyclo 2.2.2 octane, quinuclidin-3-one, chloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
102019
1-azabiciclo[2.2.2]octan-3-ona; clorhidrato
C1CN2CCC1C(=O)C2.Cl
Este artículo no se puede devolver. Vea la política de devoluciones
1193-65-3
C7H12ClNO
161.629
MFCD00137391
RFDPHKHXPMDJJD-UHFFFAOYSA-N
3-quinuclidinone hydrochloride, quinuclidin-3-one hydrochloride, 3-quinuclidone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one hydrochloride, 3-quinuclidnone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride, 1-aza-3-oxobicyclo 2.2.2 octane, quinuclidin-3-one, chloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
102019
1-azabiciclo[2.2.2]octan-3-ona; clorhidrato
C1CN2CCC1C(=O)C2.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Quinuclidinone hydrochloride
Punto de fusión ∼300°C (decomposition)
Cantidad 5 g
Beilstein 3695039
Sensibilidad Hygroscopic
Formula Weight (peso de la fórmula) 161.63
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'