missing translation for 'onlineSavingsMsg'
Learn More

3-(Fenilsulfonil)propionitrilo, 99 %, Thermo Scientific Chemicals

Catalog Number p-7048073
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
10154-75-3
C9H9NO2S
195.236
MFCD00047798
OVZPZPVSUAKTCM-UHFFFAOYSA-N
3-phenylsulfonyl propionitrile, 3-phenylsulfonyl propanenitrile, 3-phenylsulphonyl propiononitrile, 3-benzenesulfonyl propanenitrile, 3-phenylsulfonylpropionitrile, acmc-1bvrh, enamine_005271, 3-phenylsulfonyl propanitrile, 3-phenylsulphonyl-propionitrile, propanenitrile, 3-phenylsulfonyl
82413
3-(bencenosulfonil)propanonitrilo
C1=CC=C(C=C1)S(=O)(=O)CCC#N
Este artículo no se puede devolver. Vea la política de devoluciones
10154-75-3
C9H9NO2S
195.236
MFCD00047798
OVZPZPVSUAKTCM-UHFFFAOYSA-N
3-phenylsulfonyl propionitrile, 3-phenylsulfonyl propanenitrile, 3-phenylsulphonyl propiononitrile, 3-benzenesulfonyl propanenitrile, 3-phenylsulfonylpropionitrile, acmc-1bvrh, enamine_005271, 3-phenylsulfonyl propanitrile, 3-phenylsulphonyl-propionitrile, propanenitrile, 3-phenylsulfonyl
82413
3-(bencenosulfonil)propanonitrilo
C1=CC=C(C=C1)S(=O)(=O)CCC#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(Phenylsulfonyl)propionitrile
Punto de fusión 94°C to 96°C
Olor Odorless
Cantidad 5 g
Beilstein 2051019
Formula Weight (peso de la fórmula) 195.24
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'