missing translation for 'onlineSavingsMsg'
Learn More

3-O-terc-Butildimetilsilil-4,6-O-(4-metoxibencilideno)-D-glucal, 97 %, Thermo Scientific™

Catalog Number p-7040997
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
384346-91-2
C20H30O5Si
378.54
MFCD01863523
ZTRSSQRWGPRALW-UHFFFAOYNA-N
3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
71311532
COC1=CC=C(C=C1)C1OCC2OC=CC(O[Si](C)(C)C(C)(C)C)C2O1
Este artículo no se puede devolver. Vea la política de devoluciones
384346-91-2
C20H30O5Si
378.54
MFCD01863523
ZTRSSQRWGPRALW-UHFFFAOYNA-N
3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
71311532
COC1=CC=C(C=C1)C1OCC2OC=CC(O[Si](C)(C)C(C)(C)C)C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-O-tert-Butyldimethylsilyl-4,6-O-(4-methoxybenzylidene)-D-glucal
Cantidad 250 mg
Nombre IUPAC tert-butyl({[2-(4-methoxyphenyl)-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl]oxy})dimethylsilane
Formula Weight (peso de la fórmula) 378.53
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'