missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3-noradamantanocarboxílico, 98 %, Thermo Scientific Chemicals

Catalog Number p-7023287
Click to view available options
Cantidad:
0.25 g
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
16200-53-6
C10H14O2
166.22
MFCD00213463
RXUUYFUQAGICCD-NLZKFAMNSA-N
3-noradamantanecarboxylic acid, 3-noradamantane carboxylic acid, octahydro-2,5-methanopentalene-3a-carboxylic acid, 2,5-methanopentalene-3a 1h-carboxylic acid, hexahydro, 3-noradamantancarboxylic acid, 3-noradamantanecarboxylicacid, rxuuyfuqagiccd-uhfffaoysa, 3-noradamantanecarboxylate nac, hexahydro-2,5-methano-pentalene-3a-carboxylic acid, tricyclo 3.3.1.0<3,7> nonane-3-carboxylic acid
11887834
C1C2CC3CC1CC3(C2)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
16200-53-6
C10H14O2
166.22
MFCD00213463
RXUUYFUQAGICCD-NLZKFAMNSA-N
3-noradamantanecarboxylic acid, 3-noradamantane carboxylic acid, octahydro-2,5-methanopentalene-3a-carboxylic acid, 2,5-methanopentalene-3a 1h-carboxylic acid, hexahydro, 3-noradamantancarboxylic acid, 3-noradamantanecarboxylicacid, rxuuyfuqagiccd-uhfffaoysa, 3-noradamantanecarboxylate nac, hexahydro-2,5-methano-pentalene-3a-carboxylic acid, tricyclo 3.3.1.0<3,7> nonane-3-carboxylic acid
11887834
C1C2CC3CC1CC3(C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Noradamantanecarboxylic acid
Punto de fusión 105°C to 107°C
Cantidad 0.25 g
Formula Weight (peso de la fórmula) 166.22
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'