missing translation for 'onlineSavingsMsg'
Learn More

Anhídrido 3-nitroftálico, 97 %, Thermo Scientific Chemicals

Catalog Number p-7022886
Click to view available options
Cantidad:
25 g
100 g
500 g
641-70-3
C8H3NO5
193.11
MFCD00005921
ROFZMKDROVBLNY-UHFFFAOYSA-N
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
21631
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
641-70-3
C8H3NO5
193.11
MFCD00005921
ROFZMKDROVBLNY-UHFFFAOYSA-N
3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
21631
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O

4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El anhídrido 4-nitroftálico es un intermediario para la síntesis del inhibidor de PARP I de bencimidazol, ABT-472. Reacciona con aminoquinazolinonas para producir ftalimidoquinazolinonas.

Solubilidad
Puede descomponerse en agua.

Notas
Sensible al aire. Almacenar a temperatura ambiente. Manténgase lejos de los agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Nitrophthalic anhydride
Punto de fusión 161°C to 166°C
Cantidad 25 g
Beilstein 179963
Sensibilidad Moisture sensitive
Información de solubilidad May decompose in water.
Nombre IUPAC 4-nitro-1,3-dihidro-2-benzofuran-1,3-diona
Formula Weight (peso de la fórmula) 193.11
Porcentaje de pureza 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'