missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3-[N-metil-N-(2-nitrofenil)sulfamoil]bencenoborónico, 97 %, Thermo Scientific™

Catalog Number p-7035973
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1449133-23-6
C13H13BN2O6S
336.125
MFCD20265204
DXKDWPNDSRHLBL-UHFFFAOYSA-N
3-n-methyl-n-2-nitrophenyl sulfamoyl benzeneboronic acid, 3-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
73995688
ácido [3-[metil-(2-nitrofenil)sulfamoil]fenil]borónico
B(C1=CC(=CC=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O
Este artículo no se puede devolver. Vea la política de devoluciones
1449133-23-6
C13H13BN2O6S
336.125
MFCD20265204
DXKDWPNDSRHLBL-UHFFFAOYSA-N
3-n-methyl-n-2-nitrophenyl sulfamoyl benzeneboronic acid, 3-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
73995688
ácido [3-[metil-(2-nitrofenil)sulfamoil]fenil]borónico
B(C1=CC(=CC=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid
Cantidad 250 mg
Formula Weight (peso de la fórmula) 336.13
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'