Learn More
(3-Methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™
Product Code. 10712694
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712694 1 g Amber Glass Bottle 1g
10397354 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712694 Supplier Thermo Scientific Maybridge Supplier No. CC27013DA
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 154016-55-4
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD06738858
InChI Key JKVMPILAJBLISV-UHFFFAOYSA-N
Synonym 3-methylisoxazol-5-ylmethyl amine, 3-methylisoxazol-5-yl methanamine, 5-aminomethyl-3-methylisoxazole, 3-methyl-1,2-oxazol-5-yl methanamine, 3-methylisoxazol-5-yl methylamine, 1-3-methylisoxazol-5-yl methanamine, 5-isoxazolemethanamine, 3-methyl, 1-3-methyl-1,2-oxazol-5-yl methanamine, 1-3-methyl-5-isoxazolyl methanamine, c-3-methyl-isoxazol-5-yl-methylamine
PubChem CID 16481072
IUPAC Name (3-methyl-1,2-oxazol-5-yl)methanamine
SMILES CC1=NOC(=C1)CN
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (3-Methylisoxazol-5-ylmethyl)amine