Learn More
3-Metil-5-(trifluorometoxi)anilina, 97 %, Thermo Scientific™
Catalog Number p-7030543
Click to view available options
Cantidad:
250 mg
1 g
86256-63-5
C8H8F3NO
191.15
MFCD09025380
NCXWNLUZPMBTHX-UHFFFAOYSA-N
3-methyl-5-trifluoromethoxy aniline, 5-methyl-3-trifluoromethoxy phenylamine, 3-methyl-5-trifluoromethoxy aniline, jrd
46737546
3-methyl-5-(trifluoromethoxy)aniline
CC1=CC(N)=CC(OC(F)(F)F)=C1
86256-63-5
C8H8F3NO
191.15
MFCD09025380
NCXWNLUZPMBTHX-UHFFFAOYSA-N
3-methyl-5-trifluoromethoxy aniline, 5-methyl-3-trifluoromethoxy phenylamine, 3-methyl-5-trifluoromethoxy aniline, jrd
46737546
3-methyl-5-(trifluoromethoxy)aniline
CC1=CC(N)=CC(OC(F)(F)F)=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Methyl-5-(trifluoromethoxy)aniline
Punto de ebullición 73°C (1 mmHg)
Índice de refracción 1.4692
Cantidad 250 mg
Número UN UN2810
Formula Weight (peso de la fórmula) 191.15
Porcentaje de pureza 97%