Learn More
3-methoxypyridin-2-ol, TRC
Product Code. 30472257
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
30472257 250 mg 250mg
1 options
This item is not returnable. View return policy
Product Code. 30472257 Supplier Toronto Research Chemicals Supplier No. TRCM228000250MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-methoxypyridin-2-ol
CAS 20928-63-6
Molecular Formula C6H7NO2
Quantity 250 mg
InChI Formula InChI=1S/C6H7NO2/c1-9-5-3-2-4-7-6(5)8/h2-4H,1H3,(H,7,8)
SMILES COc1cccnc1O
IUPAC Name 3-methoxypyridin-2-ol
Molecular Weight (g/mol) 125.13
Formula Weight 125.0477