missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Product Code. 15495607
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15495607 1 g 1g
15405617 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15495607

Brand: Thermo Scientific Alfa Aesar H32848.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-56-4
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.137
MDL Number MFCD09025418
InChI Key DFOFNISQFZKBEY-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl-phenol, 3-methoxy-5-trifluoromethyl phenol, 3-hydroxy-5-methoxybenzotrifluoride, 3-hydroxy-5-trifluoromethyl anisole, 5-methoxy-3-trifluoromethyl phenol, 3-hydroxy-5-methoxy-benzotrifluoride, 3-methoxy-5-hydroxybenzotrifluoride, 3-hydroxy-5-methoxy-1-trifluoromethylbenzene
PubChem CID 22571708
IUPAC Name 3-methoxy-5-(trifluoromethyl)phenol
SMILES COC1=CC(=CC(=C1)O)C(F)(F)F

Specifications

Melting Point 74°C to 76°C
Quantity 1 g
Formula Weight 192.14
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.