missing translation for 'onlineSavingsMsg'
Learn More

3-Iodotoluene, 99%

Product Code. 11404453
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404453 250 g 250g
11494443 50 g 50g
11399469 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404453 Supplier Thermo Scientific Alfa Aesar Supplier No. A10999.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-95-6 | C7H7I | 218.037 g/mol

3-Iodotoluene was used in the preparartion of indene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodotoluene was used in the preparartion of indene derivatives.

Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-95-6
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001050
InChI Key VLCPISYURGTGLP-UHFFFAOYSA-N
Synonym 3-iodotoluene, m-iodotoluene, benzene, 1-iodo-3-methyl, m-tolyl iodide, m-methyliodobenzene, toluene, m-iodo, 3-methylphenyl iodide, unii-2h574ir35g, 1-iodo-3-methyl-benzene, m-iodo-toluen
PubChem CID 12268
IUPAC Name 1-iodo-3-methylbenzene
SMILES CC1=CC(=CC=C1)I

Specifications

Melting Point -27°C
Density 1.698
Boiling Point 80°C to 82°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.604
Quantity 250 g
Beilstein 1903634
Sensitivity Light sensitive
Solubility Information Not miscible in water. Soluble in benzene,alcohol and ether.
Formula Weight 218.04
Percent Purity 99%
Chemical Name or Material 3-Iodotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.