missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3-hidroxi-4-yodobenzoico, 97 %

Catalog Number 11441750
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
58123-77-6
C7H5IO3
264.02
MFCD02068386
UABBBWVTEWIIMN-UHFFFAOYSA-N
3-hydroxy-4-iodo-benzoic acid, benzoic acid, 3-hydroxy-4-iodo, acmc-1ay71, ksc497o7h, 4-iodo-3-hydroxybenzoic acid, 4-iodo-3-hydroxy-benzoic acid, 3-hydroxy-4-iodobenzoic acid, 4-iodo-3-hydroxy benzoic acid
736854
ácido 3-hidroxi-4-yodobenzoico
OC(=O)C1=CC(O)=C(I)C=C1
Este artículo no se puede devolver. Vea la política de devoluciones
58123-77-6
C7H5IO3
264.02
MFCD02068386
UABBBWVTEWIIMN-UHFFFAOYSA-N
3-hydroxy-4-iodo-benzoic acid, benzoic acid, 3-hydroxy-4-iodo, acmc-1ay71, ksc497o7h, 4-iodo-3-hydroxybenzoic acid, 4-iodo-3-hydroxy-benzoic acid, 3-hydroxy-4-iodobenzoic acid, 4-iodo-3-hydroxy benzoic acid
736854
ácido 3-hidroxi-4-yodobenzoico
OC(=O)C1=CC(O)=C(I)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Hydroxy-4-iodobenzoic acid
Punto de fusión 225°C to 231°C
Cantidad 1 g
Sensibilidad Light sensitive
Formula Weight (peso de la fórmula) 264.02
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'