Learn More
3-Formylpyrrole, TRC
Product Code. 30404978
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30404978 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30404978 Supplier Toronto Research Chemicals Supplier No. TRCF7002451G
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Formylpyrrole
CAS 7126-39-8
Molecular Formula C5H5NO
Quantity 1 g
Synonym Pyrrole-3-carboxaldehyde, 1H-Pyrrole-3-carboxaldehyde
InChI Formula InChI=1S/C5H5NO/c7-4-5-1-2-6-3-5/h1-4,6H
SMILES O=Cc1cc[nH]c1
Molecular Weight (g/mol) 95.1
Formula Weight 95.0371