missing translation for 'onlineSavingsMsg'
Learn More

Acetato de 3-fluorofenilo, 98 %, Thermo Scientific™

Catalog Number p-7046212
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
701-83-7
C8H7FO2
154.14
MFCD00142578
IAWZWMGUTKRLQB-UHFFFAOYSA-N
1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
136542
acetato de (3-fluorofenilo)
CC(=O)OC1=CC(=CC=C1)F
Este artículo no se puede devolver. Vea la política de devoluciones
701-83-7
C8H7FO2
154.14
MFCD00142578
IAWZWMGUTKRLQB-UHFFFAOYSA-N
1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
136542
acetato de (3-fluorofenilo)
CC(=O)OC1=CC(=CC=C1)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Fluorophenyl acetate
Densidad 1.15
Punto de ebullición 77°C to 78°C (13 mmHg)
Temperatura de inflamación 76°C (169°F)
Índice de refracción 1.478
Cantidad 5 g
Beilstein 2555261
Formula Weight (peso de la fórmula) 154.14
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'