missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-4'-n-propylbiphenyl-4-boronic acid, 98%

Marca:  Alfa Aesar™ H64071.03

Detalles adicionales : CAS : 909709-42-8 Peso : 0.00100kg

Cantidad. 1g

 Ver más versiones de este producto

Código de producto. 15451109

  • 68.80€ / 1 g


Fecha de envío estimada
del proveedor 14-08-2019
Añadir a la cesta

Descripción

Descripción

Applications
3-Fluoro-4′-n-propylbiphenyl-4-boronic acid is used in OLED.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Especificaciones

Identificadores de productos químicos

909709-42-8
258.099
XZGCBSYWWZAXRC-UHFFFAOYSA-N
53402576
B(C1=C(C=C(C=C1)C2=CC=C(C=C2)CCC)F)(O)O
C15H16BFO2
MFCD10687198
3-fluoro-4'-propyl-1,1'-biphenyl-4-yl boronic acid, 4'-propyl-3-fluorobiphenyl-4-boronic acid, 4-propyl-3-fluorobiphenyl-4-boronic acid, 3-fluoro-4'-propyl-biphenylboronic acid, 3-fluoro-4'-propyl-4-biphenylboronic acid, 4'-propyl-3-fluoro-4-biphenyl boronic acid, 2-fluoro-4-4-propylphenyl phenylboronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl
[2-fluoro-4-(4-propylphenyl)phenyl]boronic acid

Especificaciones

3-Fluoro-4'-n-propylbiphenyl-4-boronic acid
909709-42-8
MFCD10687198
3-fluoro-4'-propyl-1,1'-biphenyl-4-yl boronic acid, 4'-propyl-3-fluorobiphenyl-4-boronic acid, 4-propyl-3-fluorobiphenyl-4-boronic acid, 3-fluoro-4'-propyl-biphenylboronic acid, 3-fluoro-4'-propyl-4-biphenylboronic acid, 4'-propyl-3-fluoro-4-biphenyl boronic acid, 2-fluoro-4-4-propylphenyl phenylboronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl
B(C1=C(C=C(C=C1)C2=CC=C(C=C2)CCC)F)(O)O
258.099
258.1
98%
1g
C15H16BFO2
18339837
XZGCBSYWWZAXRC-UHFFFAOYSA-N
[2-fluoro-4-(4-propylphenyl)phenyl]boronic acid
53402576
Solid
Videos
SDS Documentos