missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol del ácido 3-fluoro-4-(metoxicarbonilmetil) bencenoborónico, 96 %, Thermo Scientific™

Catalog Number 15458036
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1415960-53-0
C15H20BFO4
294.129
MMANYAVWRYOHTI-UHFFFAOYSA-N
3-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 3-fluoro-4-methoxycarbonylmethyl benzeneboronicacidpinacolester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetic acid methyl ester
71265929
metil 2-[2-fluoro-4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)fenil]acetato
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC(=O)OC)F
Este artículo no se puede devolver. Vea la política de devoluciones
1415960-53-0
C15H20BFO4
294.129
MMANYAVWRYOHTI-UHFFFAOYSA-N
3-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 3-fluoro-4-methoxycarbonylmethyl benzeneboronicacidpinacolester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetic acid methyl ester
71265929
metil 2-[2-fluoro-4-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)fenil]acetato
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC(=O)OC)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester
Cantidad 250 mg
Formula Weight (peso de la fórmula) 294.13
Porcentaje de pureza 96%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'